Product Details

SNP ID
rs7933549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5200003 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTGAGAGGCAACTGCCAGGACT[A/G]ACTTAAGAATCAGGATGTAGGAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51V1 PubMed Links

Gene Details

Gene
OR51V1
Gene Name
olfactory receptor family 51 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004760.2 698 Missense Mutation TCA,TTA S233L NP_001004760.2

View Full Product Details