Product Details

SNP ID
rs34732372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140675132 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTGGTCCGTATCTTCTCCTCCAAA[A/G]CCTGGGGAAGGGGCAGATAAAAGAG
Phenotype
MIM: 609385 MIM: 142810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DND1 PubMed Links

Gene Details

Gene
DND1
Gene Name
DND microRNA-mediated repression inhibitor 1
There are no transcripts associated with this gene.

Gene
HARS
Gene Name
histidyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258040.2 1277 Missense Mutation GCT,GTT A359V NP_001244969.1
NM_001258041.2 1277 Missense Mutation GCT,GTT A379V NP_001244970.1
NM_001258042.2 1277 Missense Mutation GCT,GTT A339V NP_001244971.1
NM_001289092.1 1277 Missense Mutation GCT,GTT A325V NP_001276021.1
NM_001289093.1 1277 Missense Mutation GCT,GTT A285V NP_001276022.1
NM_001289094.1 1277 Missense Mutation GCT,GTT A370V NP_001276023.1
NM_002109.5 1277 Missense Mutation GCT,GTT A399V NP_002100.2
Gene
WDR55
Gene Name
WD repeat domain 55
There are no transcripts associated with this gene.

View Full Product Details