Product Details

SNP ID
rs8058061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23641699 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACCAACCCCTGTCCCTTTGGCC[A/G]TTAGTCCCGGATTATCTAGCGATGC
Phenotype
MIM: 612962 MIM: 610355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCTN5 PubMed Links

Gene Details

Gene
DCTN5
Gene Name
dynactin subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199011.1 Intron NP_001185940.1
NM_001199743.1 Intron NP_001186672.1
NM_032486.3 Intron NP_115875.1
Gene
PALB2
Gene Name
partner and localizer of BRCA2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024675.3 Intron NP_078951.2
XM_011545946.2 Intron XP_011544248.1
XM_011545947.2 Intron XP_011544249.1
XM_011545948.2 Intron XP_011544250.1
XM_017023671.1 Intron XP_016879160.1
XM_017023672.1 Intron XP_016879161.1
XM_017023673.1 Intron XP_016879162.1

View Full Product Details