Product Details

SNP ID
rs377384589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29912055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCGTCTCCCTTTTTTGCTGGG[G/A]AAGAAGCGGAAGGTGGTTAACAGCA
Phenotype
MIM: 608947
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ASPHD1 PubMed Links

Gene Details

Gene
ASPHD1
Gene Name
aspartate beta-hydroxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181718.3 Intron NP_859069.2
XM_017023107.1 Intron XP_016878596.1
Gene
KCTD13
Gene Name
potassium channel tetramerization domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178863.4 Intron NP_849194.1
XM_011545783.2 Intron XP_011544085.1
XM_011545784.2 Intron XP_011544086.1
XM_017023104.1 Intron XP_016878593.1
XM_017023105.1 Intron XP_016878594.1
XM_017023106.1 Intron XP_016878595.1

View Full Product Details