Product Details

SNP ID
hCV25745641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47941636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTCCGAACTCAAAGGAACCCA[A/G]TGCCGGGCCACAGCCGGGTCACGTG
Phenotype
MIM: 603287 MIM: 601801
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNPO PubMed Links

Gene Details

Gene
PNPO
Gene Name
pyridoxamine 5'-phosphate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018129.3 114 UTR 5 NP_060599.1
Gene
SP2
Gene Name
Sp2 transcription factor
There are no transcripts associated with this gene.

Gene
SP2-AS1
Gene Name
SP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details