Product Details

SNP ID
rs36102561
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:158579012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGAATATGATGGGGCTGAGGAA[A/G]GGGGTAATGACAGTATAAGGGACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10X1 PubMed Links

Gene Details

Gene
OR10X1
Gene Name
olfactory receptor family 10 subfamily X member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004477.1 888 Silent Mutation CCC,CCT P296P NP_001004477.1

View Full Product Details