Product Details

SNP ID
rs59295315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56314792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAAACCAAGGTCCTTTTTAAGCA[A/G]TCGAATTTCATCCAAAGCCAGAGTG
Phenotype
MIM: 605861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190991.1 812 UTR 3 NP_001177920.1
NM_014255.5 812 Intron NP_055070.1
Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
There are no transcripts associated with this gene.

View Full Product Details