Product Details
- SNP ID
-
hCV25756287
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:27071069 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATAAGAAATTAAGAAACTACTT[A/T]CTTCTACAATGTCGTCATCAAACAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
USP12
PubMed Links
Gene Details
- Gene
- USP12
- Gene Name
- ubiquitin specific peptidase 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182488.3 |
|
Intron |
|
|
NP_872294.2 |
View Full Product Details