Product Details
- SNP ID
-
rs11699901
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44337201 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCGTGGGCCTGGCAGGCCTGCTCT[G/T]CTGGGCTGGCCAGGCAGTGAACGCC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
R3HDML
PubMed Links
Gene Details
- Gene
- R3HDML
- Gene Name
- R3H domain containing like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178491.3 |
44 |
Missense Mutation |
TGC,TTC |
C15F |
NP_848586.1 |
View Full Product Details