Product Details
- SNP ID
-
rs1108134
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:82759440 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGAAAAACAGGGATGCACAAATT[C/T]GTGAAGCTGACAACTAAATGTATAT
- Phenotype
-
MIM: 612675
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FSD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1108135] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSD2
- Gene Name
- fibronectin type III and SPRY domain containing 2
- Gene
- SCARNA15
- Gene Name
- small Cajal body-specific RNA 15
There are no transcripts associated with this gene.
- Gene
- SNHG21
- Gene Name
- small nucleolar RNA host gene 21
There are no transcripts associated with this gene.
View Full Product Details