Product Details

SNP ID
rs72990631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139561617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATACACCTTTTGTTTAATATTGG[A/G]AAAGTGCCAAGGAATACGATTTTTT
Phenotype
MIM: 608702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507291 PubMed Links

Gene Details

Gene
LOC100507291
Gene Name
uncharacterized LOC100507291
There are no transcripts associated with this gene.

Gene
NMNAT3
Gene Name
nicotinamide nucleotide adenylyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200047.2 Intron NP_001186976.1
NM_001320510.1 Intron NP_001307439.1
NM_001320511.1 Intron NP_001307440.1
NM_001320512.1 Intron NP_001307441.1
NM_001320513.1 Intron NP_001307442.1
NM_178177.4 Intron NP_835471.1
XM_011512790.1 Intron XP_011511092.1
XM_011512791.2 Intron XP_011511093.1
XM_017006328.1 Intron XP_016861817.1
XM_017006329.1 Intron XP_016861818.1
XM_017006330.1 Intron XP_016861819.1
XM_017006331.1 Intron XP_016861820.1
XM_017006332.1 Intron XP_016861821.1
XM_017006333.1 Intron XP_016861822.1
XM_017006334.1 Intron XP_016861823.1
XM_017006335.1 Intron XP_016861824.1
XM_017006336.1 Intron XP_016861825.1
XM_017006337.1 Intron XP_016861826.1
XM_017006338.1 Intron XP_016861827.1
XM_017006339.1 Intron XP_016861828.1
XM_017006340.1 Intron XP_016861829.1
XM_017006341.1 Intron XP_016861830.1
XM_017006342.1 Intron XP_016861831.1
XM_017006343.1 Intron XP_016861832.1
XM_017006344.1 Intron XP_016861833.1

View Full Product Details