Product Details
- SNP ID
-
rs3774211
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:8621997 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AAAGTCTCGAAGTTTGAGGCCACAA[G/A]TTGCTCAAGACATCAATTGCCATCA
- Phenotype
-
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SSUH2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75073117] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SSUH2
- Gene Name
- ssu-2 homolog (C. elegans)
View Full Product Details