Product Details

SNP ID
rs34545192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22690623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCGCTGCGCGCAAACTTGCGCCC[A/G]CAGAACTCGCAGGCAAAGGGCTTCT
Phenotype
MIM: 602419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGR3 PubMed Links

Gene Details

Gene
EGR3
Gene Name
early growth response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199880.1 1323 Silent Mutation TGC,TGT C300C NP_001186809.1
NM_001199881.1 1323 Silent Mutation TGC,TGT C284C NP_001186810.1
NM_004430.2 1323 Silent Mutation TGC,TGT C338C NP_004421.2
XM_005273425.3 1323 Silent Mutation TGC,TGT C393C XP_005273482.1
XM_005273426.3 1323 Silent Mutation TGC,TGT C300C XP_005273483.1
XM_011544429.2 1323 Silent Mutation TGC,TGT C357C XP_011542731.1

View Full Product Details