Product Details

SNP ID
rs61751871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:126164967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGACTGAGAGCCCACAGCGTACA[C/G]GGACAACTCATCACAGTAGGTCAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF12L2 PubMed Links

Gene Details

Gene
DCAF12L2
Gene Name
DDB1 and CUL4 associated factor 12 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013628.2 1131 Missense Mutation CTG,GTG L320V NP_001013650.1

View Full Product Details