Product Details

SNP ID
rs12542502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144432575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAACCACTACCGCAGTGCTCG[C/T]AGGGTGGGGTGGCAAGTGCAGAGGC
Phenotype
MIM: 604546 MIM: 611952
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6893 PubMed Links

Gene Details

Gene
MIR6893
Gene Name
microRNA 6893
There are no transcripts associated with this gene.

Gene
TONSL
Gene Name
tonsoku-like, DNA repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013432.4 Intron NP_038460.4
XM_011517048.2 Intron XP_011515350.1
XM_011517049.2 Intron XP_011515351.1
XM_011517050.2 Intron XP_011515352.1
Gene
TONSL-AS1
Gene Name
TONSL antisense RNA 1
There are no transcripts associated with this gene.

Gene
VPS28
Gene Name
VPS28, ESCRT-I subunit
There are no transcripts associated with this gene.

View Full Product Details