Product Details
- SNP ID
-
rs61747967
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8324699 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCGCACGGTGTCCAGGCGCCCATA[C/T]TCACTGGCACACATCAGCGCTGTGG
- Phenotype
-
MIM: 614611
MIM: 602139
MIM: 603685
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KANK3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1019757] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KANK3
- Gene Name
- KN motif and ankyrin repeat domains 3
- Gene
- NDUFA7
- Gene Name
- NADH:ubiquinone oxidoreductase subunit A7
There are no transcripts associated with this gene.
- Gene
- RPS28
- Gene Name
- ribosomal protein S28
There are no transcripts associated with this gene.
View Full Product Details