Product Details

SNP ID
rs71539408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21501891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTCCCACCCTGGGCTTTCCGA[C/G]GTGCTGTCGCCGCTGTCCCCACCAC
Phenotype
MIM: 615078 MIM: 600537
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GOLT1B PubMed Links

Gene Details

Gene
GOLT1B
Gene Name
golgi transport 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016072.4 127 UTR 5 NP_057156.1
XM_017019384.1 127 UTR 5 XP_016874873.1
Gene
RECQL
Gene Name
RecQ like helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002907.3 127 Intron NP_002898.2
NM_032941.2 127 Intron NP_116559.1
XM_005253461.3 127 Intron XP_005253518.1
XM_005253462.4 127 Intron XP_005253519.1
XM_005253463.3 127 Intron XP_005253520.1
XM_005253464.3 127 Intron XP_005253521.1

View Full Product Details