Product Details

SNP ID
rs181567467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70260591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTAGAGGCAGTGGCTCCTCTTAA[C/T]GCCAGGCCCTAGTTGGCTCAGGCAT
Phenotype
MIM: 607448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPFFR1 PubMed Links

Gene Details

Gene
NPFFR1
Gene Name
neuropeptide FF receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022146.4 Intron NP_071429.1

View Full Product Details