Product Details

SNP ID
rs61746113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70255663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGGAGTAGAGCGGGTAGGAG[A/C]GGTTGCGGGCGTCCACCATGAAGTG
Phenotype
MIM: 607448
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NPFFR1 PubMed Links

Gene Details

Gene
NPFFR1
Gene Name
neuropeptide FF receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022146.4 605 Missense Mutation CGC,CTC R196L NP_071429.1

View Full Product Details