Product Details

SNP ID
rs113865234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73944515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGAAGTGGCCGTTAGCGAGGCT[A/G]CACACTTTGGGATCGCGTGGCCTGA
Phenotype
MIM: 300832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JPX PubMed Links

Gene Details

Gene
JPX
Gene Name
JPX transcript, XIST activator (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details