Product Details

SNP ID
rs7278128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30813009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTCAAGGGAGAGAAGATGCTAC[A/G]CCTCGGGGATTTCAGCCAATCAGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP8-1 PubMed Links

Gene Details

Gene
KRTAP8-1
Gene Name
keratin associated protein 8-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175857.3 244 UTR 3 NP_787053.1

View Full Product Details