Product Details

SNP ID
rs61743447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149765505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAATGAGGAGAAGCGGACGAGT[C/T]GGGAGAGGGCCTGGCCGGGCCGGCC
Phenotype
MIM: 614040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF467 PubMed Links

Gene Details

Gene
ZNF467
Gene Name
zinc finger protein 467
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249959.4 1110 Missense Mutation AAC,GAC N333D XP_005250016.1
XM_005249960.4 1110 Missense Mutation AAC,GAC N333D XP_005250017.1
XM_005249961.4 1110 Missense Mutation AAC,GAC N333D XP_005250018.1
XM_006715864.3 1110 Missense Mutation AAC,GAC N374D XP_006715927.1
XM_011515855.2 1110 Missense Mutation AAC,GAC N584D XP_011514157.1
XM_011515856.2 1110 Missense Mutation AAC,GAC N584D XP_011514158.1
XM_011515857.2 1110 Missense Mutation AAC,GAC N547D XP_011514159.1
XM_011515858.1 1110 Missense Mutation AAC,GAC N333D XP_011514160.1
XM_017011799.1 1110 Missense Mutation AAC,GAC N508D XP_016867288.1

View Full Product Details