Product Details

SNP ID
rs377076557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12800701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTACATAGAGTAGATAGAGTTGC[A/C]TCTGAGTTGCATCTGCAACTCTATA
Phenotype
MIM: 165161 MIM: 600538 MIM: 606034
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JUNB PubMed Links

Gene Details

Gene
JUNB
Gene Name
JunB proto-oncogene, AP-1 transcription factor subunit
There are no transcripts associated with this gene.

Gene
PRDX2
Gene Name
peroxiredoxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005809.5 Intron NP_005800.3
Gene
RNASEH2A
Gene Name
ribonuclease H2 subunit A
There are no transcripts associated with this gene.

View Full Product Details