Product Details

SNP ID
rs73801487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:20254018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAGTATGCGCTCTTCGTCTTC[C/T]CCTCTCCCCATCCGGGCCGCGCACC
Phenotype
MIM: 603746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLIT2 PubMed Links

Gene Details

Gene
SLIT2
Gene Name
slit guidance ligand 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289135.2 Intron NP_001276064.1
NM_001289136.2 Intron NP_001276065.1
NM_004787.3 Intron NP_004778.1
XM_005248211.3 Intron XP_005248268.1
XM_006713986.3 Intron XP_006714049.1
XM_011513909.2 Intron XP_011512211.1
XM_011513910.2 Intron XP_011512212.2
XM_017008845.1 Intron XP_016864334.1

View Full Product Details