Product Details

SNP ID
hCV26005335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:54000398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAACTGACAAGCACCCTTTGCAG[G/T]AAAAACCTACTGAAAAAGGCACTGT
Phenotype
MIM: 604332
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHIC2 PubMed Links

Gene Details

Gene
CHIC2
Gene Name
cysteine rich hydrophobic domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012110.3 Intron NP_036242.1

View Full Product Details