Product Details

SNP ID
rs11247703
Assay Type
Functionally Tested
NCBI dbSNP Submissions
54
Location
Chr.1:27877142 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGGGATGACAGTGAGAGGAGG[C/G]CAACAGTAAAAGGAGTCATAAGCTT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
THEMIS2 PubMed Links

Gene Details

Gene
THEMIS2
Gene Name
thymocyte selection associated family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039477.2 Intron NP_001034566.1
NM_001105556.2 Intron NP_001099026.1
NM_001286113.1 Intron NP_001273042.1
NM_001286115.1 Intron NP_001273044.1
NM_004848.3 Intron NP_004839.2
XM_005246041.3 Intron XP_005246098.1
XM_006711050.1 Intron XP_006711113.1
XM_011542445.2 Intron XP_011540747.1

View Full Product Details