Product Details

SNP ID
rs11731874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6263470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTCAGCTGGCACTGCTCAGTGA[C/T]GTAGGGGCATGACTGGGACACAGGC
Phenotype
MIM: 606201
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WFS1 PubMed Links

Gene Details

Gene
WFS1
Gene Name
wolframin ER transmembrane glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145853.1 Intron NP_001139325.1
NM_006005.3 Intron NP_005996.2
XM_017008586.1 Intron XP_016864075.1

View Full Product Details