Product Details

SNP ID
rs1054128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47403179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAGAAATTAGAGAAGGGAGGT[A/G]GGGGGGACAGGAGAGTGAAGGAATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEIS3 PubMed Links

Gene Details

Gene
MEIS3
Gene Name
Meis homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009813.2 2165 Intron NP_001009813.1
NM_001301059.1 2165 Intron NP_001287988.1
NM_020160.2 2165 Intron NP_064545.1
XM_011527136.2 2165 UTR 3 XP_011525438.1
XM_011527138.2 2165 UTR 3 XP_011525440.1
XM_011527139.2 2165 Intron XP_011525441.1
XM_011527140.1 2165 UTR 3 XP_011525442.1
XM_017027011.1 2165 UTR 3 XP_016882500.1

View Full Product Details