Product Details

SNP ID
rs3753754
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:52604721 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCAGATTTTCCCCCTTCCAAAAGAG[C/T]GGGCAGACATGCAAGTTGTGAGTTA
Phenotype
MIM: 615784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPX7 PubMed Links

Gene Details

Gene
GPX7
Gene Name
glutathione peroxidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015696.4 Intron NP_056511.2

View Full Product Details