Product Details
- SNP ID
-
rs11119587
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
57
- Location
-
Chr.1:206800370 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGGGCAGGGCAGTGGTGGTGGGGT[A/G]GGGGGGAAGGTCACAGATGAATGCT
- Phenotype
-
MIM: 605687
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL19
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114300771,rs115001401] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL19
- Gene Name
- interleukin 19
View Full Product Details