Product Details

SNP ID
rs14470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103377855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCATTTATCATGAGATTAATA[C/T]TGTGATTCCCGCTGTTTTCTTTTTC
Phenotype
MIM: 300361
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BEX3 PubMed Links

Gene Details

Gene
BEX3
Gene Name
brain expressed X-linked 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282674.1 716 UTR 3 NP_001269603.1
NM_014380.2 716 UTR 3 NP_055195.1
NM_206915.2 716 UTR 3 NP_996798.1
NM_206917.2 716 UTR 3 NP_996800.1

View Full Product Details