Product Details

SNP ID
rs2756056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103310966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGGGAGCGAGCGACGGCGGTT[A/C]TGACGCCACAACGAGGTAAGACTCA
Phenotype
MIM: 300691
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BEX2 PubMed Links

Gene Details

Gene
BEX2
Gene Name
brain expressed X-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168399.1 45 Intron NP_001161871.1
NM_001168400.1 45 Intron NP_001161872.1
NM_001168401.1 45 UTR 5 NP_001161873.1
NM_032621.3 45 UTR 5 NP_116010.1

View Full Product Details