Product Details
- SNP ID
-
rs1875909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:23443247 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCCTTTGTTGACTTTTTGAAAGAA[C/T]GATACATTGGCATAGTCCAAAACTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGA6L2
PubMed Links
Gene Details
- Gene
- GOLGA6L2
- Gene Name
- golgin A6 family-like 2
View Full Product Details