Product Details

SNP ID
rs1968675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130182517 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCCGACCTCTGCTTTCCGGGTAC[G/T]CCCGGCCCGCTGGTCGGGAGGAGCC
Phenotype
MIM: 613450 MIM: 610457
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MZT2B PubMed Links

Gene Details

Gene
MZT2B
Gene Name
mitotic spindle organizing protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263795.4 Intron XP_005263852.1
XM_011511886.2 Intron XP_011510188.1
XM_011511887.2 Intron XP_011510189.1
XM_011511888.2 Intron XP_011510190.1
XM_011511889.2 Intron XP_011510191.1
XM_011511890.2 Intron XP_011510192.1
Gene
SMPD4
Gene Name
sphingomyelin phosphodiesterase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171083.2 Intron NP_001164554.1
NM_017751.4 Intron NP_060221.2
NM_017951.4 Intron NP_060421.2
XM_011511443.2 Intron XP_011509745.1
XM_011511444.2 Intron XP_011509746.1
XM_011511445.2 Intron XP_011509747.1
XM_017004448.1 Intron XP_016859937.1
XM_017004449.1 Intron XP_016859938.1
Gene
TUBA3E
Gene Name
tubulin alpha 3e
There are no transcripts associated with this gene.

View Full Product Details