Product Details
- SNP ID
-
hCV26876353
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:49031738 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGCTCTGAATGCACAAAGTCAAC[A/C]GGGCAGATAAACCTCAAAACCAACA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM49B
PubMed Links
Gene Details
- Gene
- TRIM49B
- Gene Name
- tripartite motif containing 49B
View Full Product Details