Product Details

SNP ID
rs9703428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133165104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGGCTCAGGACCCTGTGTGAG[T/C]GGAAGCCCACCCTCTGGAAAGCCCC
Phenotype
MIM: 616237
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
KNDC1 PubMed Links

Gene Details

Gene
KNDC1
Gene Name
kinase non-catalytic C-lobe domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152643.6 Intron NP_689856.6
XM_017016858.1 Intron XP_016872347.1
XM_017016859.1 Intron XP_016872348.1

View Full Product Details