Product Details

SNP ID
rs11697472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32984769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATAATTATGGGGCCACCCACTAC[A/G]GAGACTCTACCTTTGGAGCCAGCAG
Phenotype
MIM: 613942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUN5 PubMed Links

Gene Details

Gene
SUN5
Gene Name
Sad1 and UNC84 domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080675.3 Intron NP_542406.2
XM_011528573.1 Intron XP_011526875.1
XM_011528574.1 Intron XP_011526876.1
XM_011528575.1 Intron XP_011526877.1
XM_011528576.1 Intron XP_011526878.1

View Full Product Details