Product Details

SNP ID
rs14506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127846779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGGCAGCACCTGCAGGTCTACG[T/C]TGGGGTGGAGGTGTTCGGTCCTTGC
Phenotype
MIM: 606017
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
POLR2D PubMed Links

Gene Details

Gene
POLR2D
Gene Name
RNA polymerase II subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004805.3 1812 UTR 3 NP_004796.1

View Full Product Details