Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001004298.2 | 2900 | UTR 3 | NP_001004298.2 | ||
XM_005269491.1 | 2900 | UTR 3 | XP_005269548.1 | ||
XM_005269492.2 | 2900 | UTR 3 | XP_005269549.1 | ||
XM_011539216.1 | 2900 | UTR 3 | XP_011537518.1 | ||
XM_011539217.2 | 2900 | UTR 3 | XP_011537519.1 | ||
XM_011539220.2 | 2900 | UTR 3 | XP_011537522.1 | ||
XM_011539222.2 | 2900 | UTR 3 | XP_011537524.1 | ||
XM_011539223.2 | 2900 | UTR 3 | XP_011537525.1 | ||
XM_011539224.2 | 2900 | UTR 3 | XP_011537526.1 | ||
XM_011539226.1 | 2900 | UTR 3 | XP_011537528.1 | ||
XM_011539227.2 | 2900 | UTR 3 | XP_011537529.1 | ||
XM_011539230.2 | 2900 | UTR 3 | XP_011537532.1 | ||
XM_011539231.1 | 2900 | UTR 3 | XP_011537533.1 | ||
XM_011539232.1 | 2900 | UTR 3 | XP_011537534.1 | ||
XM_011539233.1 | 2900 | UTR 3 | XP_011537535.1 | ||
XM_017015634.1 | 2900 | UTR 3 | XP_016871123.1 | ||
XM_017015635.1 | 2900 | Intron | XP_016871124.1 |