Product Details

SNP ID
rs3180493
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92487024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATATACAAAGAACAGATAATTGT[A/T]TCAGTAGGTAATAGGATATTTTATT
Phenotype
MIM: 604659 MIM: 614518 MIM: 602136
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ERVW-1 PubMed Links
Additional Information
For this assay, SNP(s) [rs79220484] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ERVW-1
Gene Name
endogenous retrovirus group W member 1
There are no transcripts associated with this gene.

Gene
GATAD1
Gene Name
GATA zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021167.4 4416 Intron NP_066990.3
Gene
PEX1
Gene Name
peroxisomal biogenesis factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000466.2 4416 UTR 3 NP_000457.1
NM_001282677.1 4416 UTR 3 NP_001269606.1
NM_001282678.1 4416 UTR 3 NP_001269607.1
XM_017012319.1 4416 Intron XP_016867808.1

View Full Product Details