Product Details

SNP ID
rs3180373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50693194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCCAAAATGAAGGCATATAGG[A/T]TCGGTCAGTTAATATTTAATTTTAA
Phenotype
MIM: 604323
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCC3 PubMed Links

Gene Details

Gene
ABCC3
Gene Name
ATP binding cassette subfamily C member 3
There are no transcripts associated with this gene.

Gene
ANKRD40
Gene Name
ankyrin repeat domain 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052855.3 4165 UTR 3 NP_443087.1
XM_017025296.1 4165 Intron XP_016880785.1

View Full Product Details