Product Details

SNP ID
rs3750066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6162888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAACATGCTGACAGAGCTGGCTG[C/T]GGCCAGTCTTGGGTGCCGGGCCAGC
Phenotype
MIM: 605081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYTH3 PubMed Links

Gene Details

Gene
CYTH3
Gene Name
cytohesin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004227.3 3360 UTR 3 NP_004218.1
Gene
USP42
Gene Name
ubiquitin specific peptidase 42
There are no transcripts associated with this gene.

View Full Product Details