Product Details

SNP ID
rs3748013
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:230322903 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ACCCAGGATGTGTGTCTTCTTCTAA[A/G]AGACCGGTTCAAAGGTGCTGGCATG
Phenotype
MIM: 616791
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGBD5 PubMed Links

Gene Details

Gene
PGBD5
Gene Name
piggyBac transposable element derived 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258311.1 3166 UTR 3 NP_001245240.1
XM_017002330.1 3166 UTR 3 XP_016857819.1

View Full Product Details