Product Details

SNP ID
rs3765809
Assay Type
Functionally Tested
NCBI dbSNP Submissions
30
Location
Chr.1:241594247 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ATGTAAAGCACCCGTTGAATTAAAA[G/T]AATTTGTTTTTGTTCAACCTCTTCC
Phenotype
MIM: 603538 MIM: 606695
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
KMO PubMed Links

Gene Details

Gene
KMO
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003679.4 1535 UTR 3 NP_003670.2
XM_005273337.1 1535 Intron XP_005273394.1
XM_005273338.3 1535 Intron XP_005273395.1
XM_017002649.1 1535 Intron XP_016858138.1
XM_017002650.1 1535 Intron XP_016858139.1
Gene
OPN3
Gene Name
opsin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014322.2 1535 UTR 3 NP_055137.2

View Full Product Details