Product Details

SNP ID
rs3769817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202214584 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCACAATATTCTTTTTAGCATTC[C/T]TAACAGGTTTTCTTAAAACAGTGTA
Phenotype
MIM: 601912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA2012 PubMed Links

Gene Details

Gene
KIAA2012
Gene Name
KIAA2012
There are no transcripts associated with this gene.

Gene
SUMO1
Gene Name
small ubiquitin-like modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005781.1 Intron NP_001005781.1
NM_001005782.1 Intron NP_001005782.1
NM_003352.4 Intron NP_003343.1

View Full Product Details