Product Details
- SNP ID
-
rs3814071
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:45900135 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAACTCAGAAGCAGGCATTGCTGA[A/C]TAGACAGTGAATTCAACTCAGTCAT
- Phenotype
-
MIM: 607293
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLIC5
PubMed Links
Gene Details
- Gene
- CLIC5
- Gene Name
- chloride intracellular channel 5
There are no transcripts associated with this gene.
View Full Product Details