Product Details

SNP ID
rs3729976
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:32334639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGGCTGGTGTAGGGCCACTCTC[T/C]GGCCCCGAGGGAGTGGATGGCTCTG
Phenotype
MIM: 601241 MIM: 602940
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
HDAC1 PubMed Links

Gene Details

Gene
HDAC1
Gene Name
histone deacetylase 1
There are no transcripts associated with this gene.

Gene
MARCKSL1
Gene Name
MARCKS like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023009.6 Intron NP_075385.1

View Full Product Details