Product Details

SNP ID
rs6803931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:16265607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGGGATTGAGTTCTAGTCTTC[C/T]GTTTGCCAGGCTTATAACATTATTA
Phenotype
MIM: 608959
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPH3 PubMed Links

Gene Details

Gene
DPH3
Gene Name
diphthamide biosynthesis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001047434.2 59 Intron NP_001040899.1
NM_206831.2 59 Intron NP_996662.1
Gene
OXNAD1
Gene Name
oxidoreductase NAD binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138381.3 59 Intron NP_612390.1
XM_005265559.3 59 UTR 5 XP_005265616.1
XM_006713405.1 59 Intron XP_006713468.1
XM_006713406.2 59 Intron XP_006713469.1
XM_011534230.2 59 UTR 5 XP_011532532.1
XM_011534232.2 59 UTR 5 XP_011532534.1
XM_011534233.2 59 UTR 5 XP_011532535.1
XM_011534234.2 59 UTR 5 XP_011532536.1
XM_011534235.2 59 UTR 5 XP_011532537.1
XM_017007485.1 59 Intron XP_016862974.1
XM_017007486.1 59 Intron XP_016862975.1
XM_017007487.1 59 Intron XP_016862976.1
XM_017007488.1 59 Intron XP_016862977.1
XM_017007489.1 59 Intron XP_016862978.1
XM_017007490.1 59 Intron XP_016862979.1
XM_017007491.1 59 Intron XP_016862980.1
XM_017007492.1 59 Intron XP_016862981.1
XM_017007493.1 59 Intron XP_016862982.1
XM_017007494.1 59 Intron XP_016862983.1
XM_017007495.1 59 Intron XP_016862984.1
XM_017007496.1 59 Intron XP_016862985.1

View Full Product Details