Product Details

SNP ID
rs113058355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:174045630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGGTGTGGGAGCGCGTTCCCC[A/G]GTAGGAGGGGGCGCGAGCGAGCAAG
Phenotype
MIM: 616752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMP19 PubMed Links

Gene Details

Gene
HMP19
Gene Name
HMP19 protein
There are no transcripts associated with this gene.

View Full Product Details