Product Details

SNP ID
rs114781878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73909222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCAAGTAATCTGGAGTCATGA[A/G]AGCTGCTAGGGGAGCAACATGAATC
Phenotype
MIM: 191840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf55 PubMed Links

Gene Details

Gene
C10orf55
Gene Name
chromosome 10 open reading frame 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001791.2 41 Intron NP_001001791.2
Gene
PLAU
Gene Name
plasminogen activator, urokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145031.2 41 Intron NP_001138503.1
NM_001319191.1 41 Intron NP_001306120.1
NM_002658.4 41 Intron NP_002649.1
XM_011539866.2 41 UTR 5 XP_011538168.1

View Full Product Details